Assay Details
| Gene Symbol: SK20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084596.2 | NP_001078065.1 | 6 - 7 | 1193 | 149 | |
| NM_001337163.1 | NP_001325355.1 | 6 - 7 | 1405 | 149 | ||
| NM_001337164.1 | NP_001325353.1 | 5 - 6 | 1144 | 149 | ||
| NM_001337165.1 | NP_001325354.1 | 5 - 6 | 1144 | 149 | ||
| NM_130158.4 | NP_566058.2 | 6 - 7 | 1341 | 149 | ||
More Information
Set Membership: |
|

SK20
Genomic Map