Assay Details
| Gene Symbol: BRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337166.1 | NP_001318432.1 | 9 - 10 | 3805 | 144 | |
| NM_001337169.1 | NP_001323761.1 | 9 - 10 | 3908 | 144 | ||
| NM_130165.3 | NP_182126.2 | 8 - 9 | 3239 | 144 | ||
More Information
Set Membership: |
|

BRM
Genomic Map