Assay Details
| Gene Symbol: MPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337177.1 | NP_001325006.1 | 5 - 6 | 1252 | 76 | |
| NM_130170.4 | NP_182131.2 | 5 - 6 | 1008 | 76 | ||
| GenBank mRNA | AK117449.1 | - | 5 - 6 | 997 | 76 | |
| BT024898.1 | - | 5 - 6 | 955 | 76 | ||
More Information
Set Membership: |
|

MPK12
Genomic Map