Assay Details
| Gene Symbol: ABIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125062.1 | NP_001118534.1 | 4 - 5 | 619 | 68 | |
| NM_001202827.1 | NP_001189756.1 | 4 - 5 | 626 | 68 | ||
| NM_130185.3 | NP_566067.1 | 4 - 5 | 619 | 68 | ||
| GenBank mRNA | AY085146.1 | - | 4 - 5 | 591 | 68 | |
| AY817012.1 | - | 4 - 5 | 424 | 68 | ||
| BT025863.1 | - | 4 - 5 | 424 | 68 | ||
More Information
Set Membership: |
|

ABIL1
Genomic Map