Assay Details
| Gene Symbol: ABIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130185.3 | NP_566067.1 | 5 - 6 | 715 | 85 | |
| GenBank mRNA | AY085146.1 | - | 5 - 6 | 687 | 85 | |
| AY817012.1 | - | 5 - 6 | 520 | 85 | ||
| BT025863.1 | - | 5 - 6 | 520 | 85 | ||
More Information
Set Membership: |
|

ABIL1
Genomic Map