Assay Details
| Gene Symbol: CPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130205.2 | NP_182164.1 | 1 - 2 | 259 | 72 | |
| GenBank mRNA | AB004871.1 | - | 1 - 2 | 231 | 72 | |
| AY074637.1 | - | 1 - 2 | 119 | 72 | ||
| AY519521.1 | - | 1 - 2 | 75 | 72 | ||
More Information
Set Membership: |
|

CPC
Genomic Map