Assay Details
| Gene Symbol: CPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130205.2 | NP_182164.1 | 2 - 3 | 352 | 104 | |
| GenBank mRNA | AB004871.1 | - | 2 - 3 | 324 | 104 | |
| AY074637.1 | - | 2 - 3 | 212 | 104 | ||
| AY519521.1 | - | 2 - 3 | 168 | 104 | ||
More Information
Set Membership: |
|

CPC
Genomic Map