Assay Details
| Gene Symbol: PRR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337212.1 | NP_001325093.1 | 2 - 3 | 612 | 84 | |
| NM_001337213.1 | NP_001325092.1 | 2 - 3 | 612 | 84 | ||
| NM_130245.5 | NP_566085.1 | 2 - 3 | 612 | 84 | ||
| GenBank mRNA | AB046953.1 | - | 2 - 3 | 345 | 84 | |
| AF272040.1 | - | 2 - 3 | 345 | 84 | ||
| AK221652.1 | - | 2 - 3 | 457 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337213.1 transcripts.
Set Membership: |
|

PRR9
Genomic Map