Assay Details
| Gene Symbol: PRR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337212.1 | NP_001325093.1 | 5 - 6 | 1137 | 103 | |
| NM_001337213.1 | NP_001325092.1 | 4 - 5 | 1224 | 103 | ||
| NM_001337214.1 | NP_001325091.1 | 4 - 5 | 1190 | 103 | ||
| NM_130245.5 | NP_566085.1 | 5 - 6 | 1137 | 103 | ||
| NM_201974.3 | NP_973703.1 | 5 - 6 | 1109 | 103 | ||
| GenBank mRNA | AB046953.1 | - | 5 - 6 | 870 | 103 | |
| AF272040.1 | - | 5 - 6 | 870 | 103 | ||
| AK221652.1 | - | 5 - 6 | 982 | 103 | ||
| AY099676.1 | - | 4 - 5 | 1192 | 103 | ||
| AY128856.1 | - | 2 - 3 | 399 | 103 | ||
More Information
Set Membership: |
|

PRR9
Genomic Map