Assay Details
| Gene Symbol: PSI-P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036474.4 | NP_001031551.1 | 3 - 4 | 476 | 72 | |
| NM_130248.5 | NP_566086.1 | 2 - 3 | 665 | 72 | ||
| GenBank mRNA | AY065156.1 | - | 2 - 3 | 421 | 72 | |
| AY114583.1 | - | 2 - 3 | 284 | 72 | ||
More Information
Set Membership: |
|

PSI-P
Genomic Map