Assay Details
| Gene Symbol: PSI-P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036474.4 | NP_001031551.1 | 5 - 6 | 621 | 75 | |
| NM_130248.5 | NP_566086.1 | 4 - 5 | 810 | 75 | ||
| GenBank mRNA | AY065156.1 | - | 4 - 5 | 566 | 75 | |
| AY114583.1 | - | 4 - 5 | 429 | 75 | ||
More Information
Set Membership: |
|

PSI-P
Genomic Map