Assay Details
| Gene Symbol: PSI-P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036474.4 | NP_001031551.1 | 6 - 7 | 659 | 107 | |
| NM_130248.5 | NP_566086.1 | 5 - 6 | 848 | 107 | ||
| GenBank mRNA | AY065156.1 | - | 5 - 6 | 604 | 107 | |
| AY114583.1 | - | 5 - 6 | 467 | 107 | ||
More Information
Set Membership: |
|

PSI-P
Genomic Map