Assay Details
| Gene Symbol: SDR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130282.2 | NP_566097.1 | 2 - 3 | 493 | 66 | |
| GenBank mRNA | AF370319.1 | - | 2 - 3 | 493 | 66 | |
| AY063106.1 | - | 1 - 2 | 464 | 66 | ||
More Information
Set Membership: |
|

SDR5
Genomic Map