Assay Details
| Gene Symbol: UPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130295.3 | NP_566098.1 | 1 | 503 | 106 | |
| GenBank mRNA | AB493596.1 | - | 1 | 224 | 106 | |
| BT024807.1 | - | 1 | 224 | 106 | ||
More Information
Set Membership: |
|

UPB1
Genomic Map