Assay Details
| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130311.2 | NP_182265.1 | 6 - 7 | 1139 | 110 | |
| GenBank mRNA | D87545.1 | - | 6 - 7 | 1139 | 110 | |
More Information
Set Membership: |
|

CKI1
Genomic Map