Assay Details
| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130311.2 | NP_182265.1 | 7 - 8 | 1522 | 79 | |
| GenBank mRNA | D87545.1 | - | 7 - 8 | 1522 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CKI1| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130311.2 | NP_182265.1 | 7 - 8 | 1522 | 79 | |
| GenBank mRNA | D87545.1 | - | 7 - 8 | 1522 | 79 | |
Set Membership: |
|