Assay Details
| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130311.2 | NP_182265.1 | 9 - 10 | 3340 | 108 | |
| GenBank mRNA | D87545.1 | - | 9 - 10 | 3340 | 108 | |
More Information
Set Membership: |
|

CKI1
Genomic Map