Assay Details
| Gene Symbol: MYB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130314.4 | NP_182268.1 | 1 - 2 | 241 | 69 | |
| GenBank mRNA | AY060588.1 | - | 1 - 2 | 193 | 69 | |
| AY142067.1 | - | 1 - 2 | 129 | 69 | ||
| AY519580.1 | - | 1 - 2 | 129 | 69 | ||
More Information
Set Membership: |
|

MYB12
Genomic Map