Assay Details
| Gene Symbol: UNE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036478.2 | NP_001031555.1 | 4 - 5 | 479 | 67 | |
| NM_001084605.1 | NP_001078074.1 | 4 - 5 | 479 | 67 | ||
| NM_130315.5 | NP_182269.1 | 4 - 5 | 645 | 67 | ||
| NM_201979.2 | NP_973708.1 | 4 - 5 | 479 | 67 | ||
| GenBank mRNA | AF083688.1 | - | 4 - 5 | 433 | 67 | |
| AJ271470.1 | - | 4 - 5 | 637 | 67 | ||
| AK316899.2 | - | 4 - 5 | 465 | 67 | ||
| AY074348.1 | - | 4 - 5 | 478 | 67 | ||
| AY091388.1 | - | 4 - 5 | 396 | 67 | ||
More Information
Set Membership: |
|

UNE5
Genomic Map