Assay Details
| Gene Symbol: UNE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036478.2 | NP_001031555.1 | 6 - 7 | 616 | 63 | |
| NM_001084605.1 | NP_001078074.1 | 6 - 7 | 616 | 63 | ||
| NM_130315.5 | NP_182269.1 | 6 - 7 | 782 | 63 | ||
| NM_201979.2 | NP_973708.1 | 6 - 7 | 616 | 63 | ||
| GenBank mRNA | AF083688.1 | - | 6 - 7 | 570 | 63 | |
| AJ271470.1 | - | 6 - 7 | 774 | 63 | ||
| AK316899.2 | - | 6 - 7 | 602 | 63 | ||
| AY074348.1 | - | 6 - 7 | 615 | 63 | ||
| AY091388.1 | - | 6 - 7 | 533 | 63 | ||
More Information
Set Membership: |
|

UNE5
Genomic Map