Assay Details
| Gene Symbol: UNE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036478.2 | NP_001031555.1 | 9 | 117 | ||
| NM_130315.5 | NP_182269.1 | 9 - 10 | 1156 | 119 | ||
| NM_201979.2 | NP_973708.1 | 9 - 10 | 995 | 119 | ||
| GenBank mRNA | AF083688.1 | - | 9 - 10 | 944 | 119 | |
| AJ271470.1 | - | 9 - 10 | 1148 | 119 | ||
| AY074348.1 | - | 9 - 10 | 989 | 119 | ||
| AY091388.1 | - | 9 - 10 | 907 | 119 | ||
More Information
Set Membership: |
|

UNE5
Genomic Map