Assay Details
| Gene Symbol: FUM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084606.2 | NP_001078075.1 | 17 - 18 | 1665 | 71 | |
| NM_001337266.1 | NP_001324328.1 | 16 - 17 | 1503 | 71 | ||
| NM_130319.4 | NP_182273.1 | 17 - 18 | 1677 | 71 | ||
| GenBank mRNA | AY054252.1 | - | 17 - 18 | 1474 | 71 | |
| AY062460.1 | - | 17 - 18 | 1518 | 71 | ||
| BT003361.1 | - | 16 - 17 | 1393 | 71 | ||
More Information
Set Membership: |
|

FUM1
Genomic Map