Assay Details
| Gene Symbol: ERF71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130320.4 | NP_182274.1 | 1 - 2 | 321 | 65 | |
| GenBank mRNA | AY074645.1 | - | 1 - 2 | 151 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF71| Gene Symbol: ERF71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130320.4 | NP_182274.1 | 1 - 2 | 321 | 65 | |
| GenBank mRNA | AY074645.1 | - | 1 - 2 | 151 | 65 | |
Set Membership: |
|