Assay Details
| Gene Symbol: DEG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125072.1 | NP_001118544.1 | 18 - 19 | 1558 | 81 | |
| NM_130361.5 | NP_566115.1 | 18 - 19 | 1611 | 81 | ||
| GenBank mRNA | AF245171.1 | - | 18 - 19 | 1542 | 81 | |
| AF326865.1 | - | 18 - 19 | 1484 | 81 | ||
| AF349516.1 | - | 18 - 19 | 1474 | 81 | ||
| AY075700.1 | - | 18 - 19 | 1484 | 81 | ||
| AY102139.1 | - | 18 - 19 | 1474 | 81 | ||
More Information
Set Membership: |
|

DEG2
Genomic Map