Assay Details
| Gene Symbol: DEG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130361.5 | NP_566115.1 | 1 - 2 | 324 | 82 | |
| GenBank mRNA | AF245171.1 | - | 1 - 2 | 255 | 82 | |
| AF326865.1 | - | 1 - 2 | 197 | 82 | ||
| AF349516.1 | - | 1 - 2 | 187 | 82 | ||
| AY075700.1 | - | 1 - 2 | 197 | 82 | ||
| AY102139.1 | - | 1 - 2 | 187 | 82 | ||
More Information
Set Membership: |
|

DEG2
Genomic Map