Assay Details
| Gene Symbol: ACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100683.3 | NP_172287.1 | 2 - 3 | 384 | 156 | |
| GenBank mRNA | AY122907.1 | - | 2 - 3 | 319 | 156 | |
More Information
Set Membership: |
|

ACA7
Genomic Map