Assay Details
| Gene Symbol: ACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100683.3 | NP_172287.1 | 3 - 4 | 497 | 71 | |
| GenBank mRNA | AY122907.1 | - | 3 - 4 | 432 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACA7| Gene Symbol: ACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100683.3 | NP_172287.1 | 3 - 4 | 497 | 71 | |
| GenBank mRNA | AY122907.1 | - | 3 - 4 | 432 | 71 | |
Set Membership: |
|