Assay Details
| Gene Symbol: ACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100683.3 | NP_172287.1 | 5 - 6 | 766 | 63 | |
| GenBank mRNA | AY122907.1 | - | 5 - 6 | 701 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACA7| Gene Symbol: ACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100683.3 | NP_172287.1 | 5 - 6 | 766 | 63 | |
| GenBank mRNA | AY122907.1 | - | 5 - 6 | 701 | 63 | |
Set Membership: |
|