Assay Details
| Gene Symbol: AT1G08340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100707.4 | NP_172310.1 | 2 - 3 | 783 | 116 | |
| GenBank mRNA | AK228173.1 | - | 2 - 3 | 606 | 116 | |
| BT012608.1 | - | 2 - 3 | 552 | 116 | ||
| BT014961.1 | - | 2 - 3 | 366 | 116 | ||
More Information
Set Membership: |
|

AT1G08340
Genomic Map