Assay Details
| Gene Symbol: HDA08 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331774.1 | NP_001320319.1 | 1 - 2 | 574 | 72 | |
| NM_100719.4 | NP_563817.1 | 1 - 2 | 574 | 72 | ||
| GenBank mRNA | AF410272.1 | - | 1 - 2 | 309 | 72 | |
| AF510167.1 | - | 1 - 2 | 245 | 72 | ||
| AY097371.1 | - | 1 - 2 | 245 | 72 | ||
More Information
Set Membership: |
|

HDA08
Genomic Map