Assay Details
| Gene Symbol: HDA08 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100719.4 | NP_563817.1 | 2 - 3 | 1160 | 62 | |
| GenBank mRNA | AF410272.1 | - | 2 - 3 | 895 | 62 | |
| AF510167.1 | - | 2 - 3 | 831 | 62 | ||
| AY097371.1 | - | 2 - 3 | 831 | 62 | ||
More Information
Set Membership: |
|

HDA08
Genomic Map