Assay Details
| Gene Symbol: AT1G08760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100750.3 | NP_172353.1 | 2 - 3 | 1720 | 68 | |
| GenBank mRNA | AY099633.1 | - | 2 - 3 | 1393 | 68 | |
| BT010341.1 | - | 2 - 3 | 1286 | 68 | ||
More Information
Set Membership: |
|

AT1G08760
Genomic Map