Assay Details
| Gene Symbol: MYB60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331790.1 | NP_001318958.1 | 1 - 2 | 285 | 70 | |
| NM_100755.3 | NP_172358.1 | 1 - 2 | 285 | 70 | ||
| GenBank mRNA | AK117469.1 | - | 1 - 2 | 155 | 70 | |
| AY519551.1 | - | 1 - 2 | 136 | 70 | ||
| BT005074.1 | - | 1 - 2 | 136 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001331790.1 transcripts.
Set Membership: |
|

MYB60
Genomic Map