Assay Details
| Gene Symbol: CSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084025.1 | NP_001077494.1 | 3 - 4 | 266 | 65 | |
| NM_100757.4 | NP_172360.1 | 3 - 4 | 333 | 65 | ||
| GenBank mRNA | AY050932.1 | - | 3 - 4 | 276 | 65 | |
| AY087273.1 | - | 3 - 4 | 307 | 65 | ||
| AY091168.1 | - | 2 - 3 | 181 | 65 | ||
| X60935.1 | - | 293 | 65 | |||
More Information
Set Membership: |
|

CSD1
Genomic Map