Assay Details
| Gene Symbol: NP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331801.1 | NP_001323260.1 | 11 - 12 | 1137 | 92 | |
| NM_100771.4 | NP_563832.2 | 11 - 12 | 1135 | 92 | ||
| GenBank mRNA | AK117282.1 | - | 11 - 12 | 1119 | 92 | |
| BT005949.1 | - | 11 - 12 | 1045 | 92 | ||
More Information
Set Membership: |
|

NP1
Genomic Map