Assay Details
| Gene Symbol: NP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331801.1 | NP_001323260.1 | 13 - 14 | 1408 | 144 | |
| NM_100771.4 | NP_563832.2 | 13 - 14 | 1406 | 144 | ||
| GenBank mRNA | AK117282.1 | - | 13 - 14 | 1390 | 144 | |
| BT005949.1 | - | 13 - 14 | 1316 | 144 | ||
More Information
Set Membership: |
|

NP1
Genomic Map