Assay Details
| Gene Symbol: NP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100771.4 | NP_563832.2 | 15 - 16 | 1925 | 76 | |
| GenBank mRNA | AK117282.1 | - | 15 - 16 | 1909 | 76 | |
| BT005949.1 | - | 15 - 16 | 1835 | 76 | ||
More Information
Set Membership: |
|

NP1
Genomic Map