Assay Details
| Gene Symbol: NP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331801.1 | NP_001323260.1 | 1 - 2 | 388 | 96 | |
| NM_100771.4 | NP_563832.2 | 1 - 2 | 386 | 96 | ||
| GenBank mRNA | AK117282.1 | - | 1 - 2 | 370 | 96 | |
| BT005949.1 | - | 1 - 2 | 296 | 96 | ||
More Information
Set Membership: |
|

NP1
Genomic Map