Assay Details
| Gene Symbol: NF-YB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100774.2 | NP_172377.1 | 1 | 389 | 94 | |
| GenBank mRNA | BT029363.1 | - | 1 | 387 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF-YB4| Gene Symbol: NF-YB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100774.2 | NP_172377.1 | 1 | 389 | 94 | |
| GenBank mRNA | BT029363.1 | - | 1 | 387 | 94 | |
Set Membership: |
|