Assay Details
| Gene Symbol: RPT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100781.5 | NP_172384.1 | 1 - 2 | 224 | 132 | |
| GenBank mRNA | AF412095.1 | - | 1 - 2 | 148 | 132 | |
| AY050423.1 | - | 1 - 2 | 148 | 132 | ||
| AY124813.1 | - | 1 - 2 | 119 | 132 | ||
More Information
Set Membership: |
|

RPT5B
Genomic Map