Assay Details
| Gene Symbol: CRT1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100791.4 | NP_172392.1 | 12 - 13 | 1607 | 95 | |
| GenBank mRNA | AY045656.1 | - | 12 - 13 | 1302 | 95 | |
More Information
Set Membership: |
|

CRT1b
Genomic Map