Assay Details
| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331830.1 | NP_001322308.1 | 4 - 5 | 985 | 72 | |
| NM_100804.4 | NP_172405.1 | 4 - 5 | 645 | 72 | ||
| GenBank mRNA | AF325043.2 | - | 4 - 5 | 417 | 72 | |
| AF428282.1 | - | 4 - 5 | 430 | 72 | ||
| AY035050.1 | - | 4 - 5 | 487 | 72 | ||
| AY070022.1 | - | 4 - 5 | 413 | 72 | ||
More Information
Set Membership: |
|

CRB
Genomic Map