Assay Details
| Gene Symbol: G6PD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198018.1 | NP_001184947.1 | 7 - 8 | 1227 | 124 | |
| NM_100813.2 | NP_563844.1 | 7 - 8 | 1173 | 124 | ||
| GenBank mRNA | AK229085.1 | - | 7 - 8 | 1190 | 124 | |
| AY056232.1 | - | 7 - 8 | 1158 | 124 | ||
| AY117271.1 | - | 7 - 8 | 1069 | 124 | ||
More Information
Set Membership: |
|

G6PD4
Genomic Map