Assay Details
| Gene Symbol: ACLA-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100814.4 | NP_172414.1 | 7 - 8 | 902 | 94 | |
| GenBank mRNA | AY127019.1 | - | 7 - 8 | 794 | 94 | |
| BT003017.1 | - | 7 - 8 | 763 | 94 | ||
More Information
Set Membership: |
|

ACLA-3
Genomic Map