Assay Details
| Gene Symbol: Haspin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100816.5 | NP_172416.2 | 7 - 8 | 1175 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Haspin| Gene Symbol: Haspin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100816.5 | NP_172416.2 | 7 - 8 | 1175 | 153 | |
Set Membership: |
|