Assay Details
| Gene Symbol: CDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100849.3 | NP_172448.1 | 1 - 2 | 354 | 112 | |
| GenBank mRNA | AY093057.1 | - | 1 - 2 | 345 | 112 | |
| BT008801.1 | - | 1 - 2 | 184 | 112 | ||
More Information
Set Membership: |
|

CDC5
Genomic Map