Assay Details
| Gene Symbol: CDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100849.3 | NP_172448.1 | 2 - 3 | 2073 | 93 | |
| GenBank mRNA | AY093057.1 | - | 2 - 3 | 2064 | 93 | |
| BT008801.1 | - | 2 - 3 | 1903 | 93 | ||
More Information
Set Membership: |
|

CDC5
Genomic Map