Assay Details
| Gene Symbol: POLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100855.4 | NP_563854.1 | 1 - 2 | 336 | 71 | |
| GenBank mRNA | BT025596.1 | - | 1 - 2 | 177 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLD4| Gene Symbol: POLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100855.4 | NP_563854.1 | 1 - 2 | 336 | 71 | |
| GenBank mRNA | BT025596.1 | - | 1 - 2 | 177 | 71 | |
Set Membership: |
|