Assay Details
| Gene Symbol: RAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100869.4 | NP_172466.1 | 1 | 1125 | 139 | |
| GenBank mRNA | AK228972.1 | - | 1 | 836 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAS1| Gene Symbol: RAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100869.4 | NP_172466.1 | 1 | 1125 | 139 | |
| GenBank mRNA | AK228972.1 | - | 1 | 836 | 139 | |
Set Membership: |
|