Assay Details
| Gene Symbol: ERG28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100877.3 | NP_563858.1 | 1 - 2 | 335 | 65 | |
| GenBank mRNA | AK227737.1 | - | 1 - 2 | 206 | 65 | |
| BT008773.1 | - | 1 - 2 | 116 | 65 | ||
More Information
Set Membership: |
|

ERG28
Genomic Map